Learn More
Waters Corp WATERS CORPORATION Column, C18, 3.5 μm, 4.6 mm × 20 mm
Supplier:  Waters Corp 186003029
Encompass_Preferred
The BEH C18 sorbent is a robust solution for HPLC method development, compatible with mobile phase pH ranging from 1 to 12. Its trifunctionally bonded C18 ligand ensures a universal column for most HPLC separations, delivering the widest usable pH range, superior low pH stability, and ultra-low column bleed.
  • Workhorse for HPLC method development
  • Usable over an entire range of mobile phase pH (1 to 12)
  • Trifunctionally bonded C18 ligand
  • Universal column for most HPLC separations
  • Widest usable pH range
  • Superior low pH stability
  • Ultra-low column bleed
Catalog No. 50-781-482