Learn More
Waters Corp HPLC Column, C18, 5.0 μm, 4.6 mm ID, 250 mm L
Supplier:  Waters Corp 186003776
Encompass_Preferred
This HPLC column is designed as a workhorse for method development, offering versatility across a wide range of mobile phase pH values. Its trifunctionally bonded C18 ligand provides a universal column suitable for most HPLC separations. The column is characterized by superior stability in low pH conditions and minimal column bleed.
  • Can be used over an entire range of mobile phase pH (1 to 12).
  • Trifunctionally bonded C18 ligand provides a universal column for most HPLC separations.
  • Offers the widest usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed.
Catalog No. 50-465-957