Learn More
Waters Corp XBridge C8 OSM Cartridge, 10 μm, 1 × 10 mm, 96 pk
Catalog No. 502392121 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge C8 OSM Solid-Phase Extraction (SPE) Cartridges are designed for use with the ACQUITY UPLC Online SPE Manager (OSM). Featuring Ethylene Bridged Hybrid (BEH) Technology, the C8 sorbent offers high mechanical strength, excellent peak shape, efficiency, and pH stability (pH 1–12). These 1 × 10 mm narrow bore PEEK cartridges can withstand up to 15,000 psi (1,000 bar) and come in trays of 96, intended for single use.
  • High mechanical strength from BEH Technology.
  • Excellent peak shape and high efficiency.
  • Superior pH stability (pH 1–12).
  • Pressure tolerance up to 15,000 psi (1,000 bar).
  • Compatible with ACQUITY UPLC Online SPE Manager for automated SPE.
  • Wide solvent compatibility.
  • Less retentive than C18 sorbent.
  • Single-use design ensures consistent performance.
Catalog No. 50-239-2121 Supplier Waters Corp Supplier No. 186005673
XBridge C8 OSM Solid-Phase Extraction (SPE) Cartridges are designed for use with the ACQUITY UPLC Online SPE Manager (OSM). Featuring Ethylene Bridged Hybrid (BEH) Technology, the C8 sorbent offers high mechanical strength, excellent peak shape, efficiency, and pH stability (pH 1–12). These 1 × 10 mm narrow bore PEEK cartridges can withstand up to 15,000 psi (1,000 bar) and come in trays of 96, intended for single use.
  • High mechanical strength from BEH Technology.
  • Excellent peak shape and high efficiency.
  • Superior pH stability (pH 1–12).
  • Pressure tolerance up to 15,000 psi (1,000 bar).
  • Compatible with ACQUITY UPLC Online SPE Manager for automated SPE.
  • Wide solvent compatibility.
  • Less retentive than C18 sorbent.
  • Single-use design ensures consistent performance.