Learn More
Waters Corp Waters XBridge BEH C8 Column, 3.5 μm, 130 Å, 1 mm × 100 mm
Catalog No. 50781606 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent. It is suitable for methods where speed is preferred, while also improving peak shape and chromatographic performance. This trifunctionally bonded sorbent offers the widest usable pH range (1–12), superior low pH stability, and ultra-low column bleed.
  • Seamless method migration to UltraPerformance LC technology is enabled by BEH Technology.
  • Exceptional chemical and mechanical stability ensures long column lifetimes under harsh operating conditions.
  • A single column handles both low and high pH conditions, reducing downtime and cost.
  • Manufactured in cGMP, ISO9001, and ISO13485 certified facilities.
Catalog No. 50-781-606 Supplier Waters Corp Supplier No. 186003178
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent. It is suitable for methods where speed is preferred, while also improving peak shape and chromatographic performance. This trifunctionally bonded sorbent offers the widest usable pH range (1–12), superior low pH stability, and ultra-low column bleed.
  • Seamless method migration to UltraPerformance LC technology is enabled by BEH Technology.
  • Exceptional chemical and mechanical stability ensures long column lifetimes under harsh operating conditions.
  • A single column handles both low and high pH conditions, reducing downtime and cost.
  • Manufactured in cGMP, ISO9001, and ISO13485 certified facilities.