Learn More
Waters Corp Waters Column, C8, 3.5 μm, 130 Å, 3 mm × 150 mm
Catalog No. 50781505 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C8 column offers less retention than C18 sorbents, improving peak shape and chromatographic performance. Its trifunctionally bonded C8 sorbent provides a wide pH range (1 to 12), superior low pH stability, and ultra-low column bleed. It ensures reliability, robustness, extended lifespan, and reproducible results for LC/MS and QC methods.
  • Provides less retention than C18 sorbents for faster analysis and improved peak shape.
  • Trifunctionally bonded C8 sorbent offers a wide pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • Ensures better column reliability, assay robustness, and a lengthy column lifetime.
  • Maximizes particle efficiency, delivering unrivaled peak form and consistent, reproducible results.
  • Rigorously tested from raw material to final batch for quality assurance.
Catalog No. 50-781-505 Supplier Waters Corp Supplier No. 186003052
The XBridge BEH C8 column offers less retention than C18 sorbents, improving peak shape and chromatographic performance. Its trifunctionally bonded C8 sorbent provides a wide pH range (1 to 12), superior low pH stability, and ultra-low column bleed. It ensures reliability, robustness, extended lifespan, and reproducible results for LC/MS and QC methods.
  • Provides less retention than C18 sorbents for faster analysis and improved peak shape.
  • Trifunctionally bonded C8 sorbent offers a wide pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • Ensures better column reliability, assay robustness, and a lengthy column lifetime.
  • Maximizes particle efficiency, delivering unrivaled peak form and consistent, reproducible results.
  • Rigorously tested from raw material to final batch for quality assurance.