Learn More
Waters Corp Waters XBridge BEH C8 Column, 3.5 μm, 4.6 mm ID, 100 mm Length
Catalog No. 50781507 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent, offering speed, improved peak shape, and chromatographic performance. The trifunctionally bonded sorbent provides the widest usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • Less retentive compared to BEH C18 sorbent.
  • Offers the widest usable pH range (1 to 12).
  • Superior low pH stability.
  • Ultra-low column bleed.
Catalog No. 50-781-507 Supplier Waters Corp Supplier No. 186003054
Add to Cart
Add to Cart
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent, offering speed, improved peak shape, and chromatographic performance. The trifunctionally bonded sorbent provides the widest usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • Less retentive compared to BEH C18 sorbent.
  • Offers the widest usable pH range (1 to 12).
  • Superior low pH stability.
  • Ultra-low column bleed.