Learn More
Waters Corp Waters XBridge BEH C8 Column, 5 μm, 2.1 mm x 150 mm
Supplier:  Waters Corp 186003013
Encompass_Preferred
The XBridge BEH C8 sorbent is characterized by being less retentive compared to the XBridge BEH C18 sorbent, which can be beneficial for faster method development. It aims to improve peak shape and chromatographic performance.
  • Less retentive compared to BEH C18 sorbent.
  • Improves peak shape and chromatographic performance.
  • Trifunctionally bonded sorbent.
  • Offers the widest usable pH range (1 to 12).
  • Superior low pH stability.
  • Ultra-low column bleed.
Catalog No. 50-781-466