Learn More
Waters Corp Column, XBridge BEH C8, 130Å, 5 μm, 2.1 mm X 50 mm
Supplier:  Waters Corp 186003011
Encompass_Preferred
The XBridge BEH C8 column offers faster method development and enhanced chromatographic performance due to its less retentive C8 sorbent. Utilizing Ethylene Bridged Hybrid (BEH) Particle Technology, it provides a wide usable pH range of 1 to 12, superior low pH stability, and ultra-low column bleed. This design ensures sustained column performance, extended lifetime, and symmetrical peak shapes by minimizing adverse interactions.
  • Less retentive C8 sorbent for faster method development.
  • Improves peak shape and chromatographic performance.
  • Trifunctionally bonded sorbent with ultra-low column bleed.
  • Wide usable pH range of 1 to 12.
  • Superior low pH stability.
  • Ethylene Bridged Hybrid (BEH) particle technology for extended lifetime.
  • Ensures high efficiencies and symmetrical peak shapes.
  • Minimizes adverse interactions by reducing free silanol sites.
Catalog No. 50-781-464