Learn More
Waters Corp Waters Column, C8, 130Å, 5 μm, 3 mm × 150 mm
Catalog No. 50781467 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C8 Column is designed for reversed-phase HPLC separations, offering excellent pH stability and long column lifetime. It utilizes Ethylene Bridged Hybrid (BEH) Technology and C8 chemistry for efficient and robust chromatographic performance.
  • Less retentive compared to C18 sorbent.
  • Trifunctionally bonded sorbent provides the widest usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • Improved pH stability over a wide pH range, high efficiency, and symmetrical peak shape.
  • Utilizes Ethylene Bridged Hybrid (BEH) Technology for high column performance and a long column lifetime in hostile environments.
  • BEH particle dependency enhances particle strength and decreases free silanol sites to reduce negative interactions with injected material, leading to improved peak shape.
  • Can operate at temperatures up to 60°C.
Catalog No. 50-781-467 Supplier Waters Corp Supplier No. 186003014
The XBridge BEH C8 Column is designed for reversed-phase HPLC separations, offering excellent pH stability and long column lifetime. It utilizes Ethylene Bridged Hybrid (BEH) Technology and C8 chemistry for efficient and robust chromatographic performance.
  • Less retentive compared to C18 sorbent.
  • Trifunctionally bonded sorbent provides the widest usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • Improved pH stability over a wide pH range, high efficiency, and symmetrical peak shape.
  • Utilizes Ethylene Bridged Hybrid (BEH) Technology for high column performance and a long column lifetime in hostile environments.
  • BEH particle dependency enhances particle strength and decreases free silanol sites to reduce negative interactions with injected material, leading to improved peak shape.
  • Can operate at temperatures up to 60°C.