Learn More
Waters Corp HPLC Column, BEH C8, 5 μm, 3 mm x 50 mm
Supplier:  Waters Corp 186003190
Encompass_Preferred
The XBridge BEH C8 Column is designed for flexible HPLC methods, enabling robust and confident separations. It operates across a wide pH and temperature range, combining advanced bonding, endcapping, and particle synthesis for challenging chromatographic separations.
  • Operates across a wide mobile phase pH (1-12) and temperature range.
  • Facilitates method transfer to UPLC technology.
  • Offers unmatched column lifetime with robust design.
  • Features BEH Technology™ for high pH dissolution and structural integrity.
  • Reduces USP peak tailing for basic compounds.
  • Enhances chemical and mechanical stability.
  • Delivers excellent reproducibility and performance.
  • Allows scaling between UPLC, analytical, and preparative platforms.
Catalog No. 50-781-618