Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters XBridge BEH C8 Column, 130 Å, 5 μm, 4.6 mm X 100 mm

Supplier:  Waters Corp 186003016

Encompass_Preferred

The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent. It is preferred for method development due to the speed offered by a less retentive column, while also improving peak shape and chromatographic performance. This trifunctionally bonded sorbent provides the widest usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.

  • Less retentive C8 sorbent for faster method development and improved peak shape
  • Trifunctionally bonded sorbent
  • Widest usable pH range (1 to 12)
  • Superior low pH stability
  • Ultra-low column bleed
  • Separation mode: Reversed Phase
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • Suitable for HPLC systems
  • BEH particle technology

Catalog No. 50-781-469


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.