Learn More
Waters Corp Waters XBridge BEH C8 Column, 5 μm, 4.6 mm × 250 mm
Catalog No. 50781471 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C8 column offers less retention than C18, improving peak shape and chromatographic performance for faster method development across a wide pH range (1 to 12).
  • Trifunctionally bonded sorbent provides superior low pH stability and ultra-low column bleed.
  • Offers unmatched potential column lifetime and flexibility across mobile phase pH and temperatures.
  • Enables robust separations and seamless migration to UPLC technology.
  • Proprietary BEH Technology facilitates scaling across UPLC, analytical, and preparative platforms.
  • Features reduced USP peak tailing, increased chemical/mechanical stability, and high pH stability.
  • Each batch is chromatographically tested for excellent, reproducible performance.
  • Manufactured in a cGMP, ISO 9001:2000 certified plant using ultra-pure reagents.
Catalog No. 50-781-471 Supplier Waters Corp Supplier No. 186003018
Add to Cart
Add to Cart
The XBridge BEH C8 column offers less retention than C18, improving peak shape and chromatographic performance for faster method development across a wide pH range (1 to 12).
  • Trifunctionally bonded sorbent provides superior low pH stability and ultra-low column bleed.
  • Offers unmatched potential column lifetime and flexibility across mobile phase pH and temperatures.
  • Enables robust separations and seamless migration to UPLC technology.
  • Proprietary BEH Technology facilitates scaling across UPLC, analytical, and preparative platforms.
  • Features reduced USP peak tailing, increased chemical/mechanical stability, and high pH stability.
  • Each batch is chromatographically tested for excellent, reproducible performance.
  • Manufactured in a cGMP, ISO 9001:2000 certified plant using ultra-pure reagents.