Learn More
Waters Corp XBridge BEH C8 Column, 130Å, 5 µm, 4.6 mm X 30 mm, 1/pk
Catalog No. 50781622 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent. Anyone developing methods may prefer the speed offered by a less retentive column, while improving peak shape and chromatographic performance. The trifunctionally bonded sorbent offers you the widest usable pH range (1-12), superior low pH stability, and ultra-low column bleed.
Catalog No. 50-781-622 Supplier Waters Corp Supplier No. 186003194
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent. Anyone developing methods may prefer the speed offered by a less retentive column, while improving peak shape and chromatographic performance. The trifunctionally bonded sorbent offers you the widest usable pH range (1-12), superior low pH stability, and ultra-low column bleed.