Learn More
Waters Corp HPLC Guard Cartridge, BEH C8, 130Å, 5 μm
Catalog No. 50781533 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This HPLC guard cartridge is designed for use in High-Performance Liquid Chromatography systems. It features a BEH C8 stationary phase, a pore size of 130Å, and a particle size of 5 μm, with dimensions of 3 mm x 20 mm.
  • Designed for use in high-performance liquid chromatography systems.
  • Features a BEH C8 stationary phase.
  • Has a pore size of 130Å.
  • Has a particle size of 5 μm.
  • Dimensions are 3 mm x 20 mm.
  • Available in a pack of two.
Catalog No. 50-781-533 Supplier Waters Corp Supplier No. 186003081
This HPLC guard cartridge is designed for use in High-Performance Liquid Chromatography systems. It features a BEH C8 stationary phase, a pore size of 130Å, and a particle size of 5 μm, with dimensions of 3 mm x 20 mm.
  • Designed for use in high-performance liquid chromatography systems.
  • Features a BEH C8 stationary phase.
  • Has a pore size of 130Å.
  • Has a particle size of 5 μm.
  • Dimensions are 3 mm x 20 mm.
  • Available in a pack of two.