Learn More
Waters Corp Waters Column, C8, Intelligent Speed (IS), 3.5 μm, 2.1 mm x 20 mm, 130 Å
Catalog No. 50781608 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent, which can be preferred by those developing methods for its speed, improved peak shape, and chromatographic performance.
  • Trifunctionally bonded sorbent provides the widest usable pH range (1 to 12).
  • Superior low pH stability.
  • Ultra-low column bleed.
Catalog No. 50-781-608 Supplier Waters Corp Supplier No. 186003180
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent, which can be preferred by those developing methods for its speed, improved peak shape, and chromatographic performance.
  • Trifunctionally bonded sorbent provides the widest usable pH range (1 to 12).
  • Superior low pH stability.
  • Ultra-low column bleed.