Learn More
Waters Corp Waters Column, C8, 3.5 μm, 3 mm x 20 mm, Intelligent Speed
Catalog No. 50781609 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent. It is preferred for method development due to the speed offered by a less retentive column, while improving peak shape and chromatographic performance. The trifunctionally bonded sorbent offers the widest usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • Less retentive compared to the C18 sorbent.
  • Offers speed for method development.
  • Improves peak shape and chromatographic performance.
  • Trifunctionally bonded sorbent provides the widest usable pH range (1 to 12).
  • Superior low pH stability.
  • Ultra-low column bleed.
Catalog No. 50-781-609 Supplier Waters Corp Supplier No. 186003181
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent. It is preferred for method development due to the speed offered by a less retentive column, while improving peak shape and chromatographic performance. The trifunctionally bonded sorbent offers the widest usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • Less retentive compared to the C18 sorbent.
  • Offers speed for method development.
  • Improves peak shape and chromatographic performance.
  • Trifunctionally bonded sorbent provides the widest usable pH range (1 to 12).
  • Superior low pH stability.
  • Ultra-low column bleed.