Learn More
Waters Corp Waters Method Validation Kit, C8, 3.5 μm, 2.1 mm x 100 mm
Catalog No. 50465958 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit features a C8 sorbent designed for reversed-phase separation. It offers less retention compared to C18 sorbents, providing faster analysis and improved chromatographic performance. The trifunctionally bonded sorbent ensures a broad pH range of 1 to 12, superior low pH stability, and ultra-low column bleed.
  • C8 chemistry for reversed-phase separation
  • Hybrid particles; 3.5 μm particle size, 130 Å pore size
  • Broad pH range from 1 to 12
  • Maximum pressure of 6000 psi (415 bar)
  • 2.1 mm internal diameter, 100 mm length
  • Method validation kit; 3 units per package
  • Designed for faster analysis and improved peak shape
Catalog No. 50-465-958 Supplier Waters Corp Supplier No. 186003777
This method validation kit features a C8 sorbent designed for reversed-phase separation. It offers less retention compared to C18 sorbents, providing faster analysis and improved chromatographic performance. The trifunctionally bonded sorbent ensures a broad pH range of 1 to 12, superior low pH stability, and ultra-low column bleed.
  • C8 chemistry for reversed-phase separation
  • Hybrid particles; 3.5 μm particle size, 130 Å pore size
  • Broad pH range from 1 to 12
  • Maximum pressure of 6000 psi (415 bar)
  • 2.1 mm internal diameter, 100 mm length
  • Method validation kit; 3 units per package
  • Designed for faster analysis and improved peak shape