Learn More
Waters Corp XBridge BEH C8 Method Validation Kit, 130Å, 3.5 μm, 3 mm x 150 mm
Catalog No. 50465960 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent. It is suitable for method development, offering speed while improving peak shape and chromatographic performance. The trifunctionally bonded sorbent provides the widest usable pH range (1-12), superior low pH stability, and ultra-low column bleed.
  • Less retentive compared to C18 sorbents.
  • Suitable for method development, improving peak shape and chromatographic performance.
  • Trifunctionally bonded sorbent.
  • Widest usable pH range (1 to 12).
  • Superior low pH stability.
  • Ultra-low column bleed.
Catalog No. 50-465-960 Supplier Waters Corp Supplier No. 186003779
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent. It is suitable for method development, offering speed while improving peak shape and chromatographic performance. The trifunctionally bonded sorbent provides the widest usable pH range (1-12), superior low pH stability, and ultra-low column bleed.
  • Less retentive compared to C18 sorbents.
  • Suitable for method development, improving peak shape and chromatographic performance.
  • Trifunctionally bonded sorbent.
  • Widest usable pH range (1 to 12).
  • Superior low pH stability.
  • Ultra-low column bleed.