Learn More
Waters Corp XBridge BEH C8 Method Validation Kit, 130Å, 5 μm, 3 mm × 100 mm
Catalog No. 50465964 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XBridge BEH C8 Method Validation Kit is designed for method development and validation, offering reproducibility and flexibility with three columns from different batches.
  • Built with tri-functionally bonded BEH particles
  • Extended column lifetime
  • Wide pH range (pH 1 to 12) suitable for various chromatographic separations
  • C8 ligand is less retentive than C18
  • Ideal for hydrophobic and extremely hydrophobic compounds
Catalog No. 50-465-964 Supplier Waters Corp Supplier No. 186003783
This XBridge BEH C8 Method Validation Kit is designed for method development and validation, offering reproducibility and flexibility with three columns from different batches.
  • Built with tri-functionally bonded BEH particles
  • Extended column lifetime
  • Wide pH range (pH 1 to 12) suitable for various chromatographic separations
  • C8 ligand is less retentive than C18
  • Ideal for hydrophobic and extremely hydrophobic compounds