Learn More
Waters Corp XBridge Column, Amide, 2.5 μm, 130 Å, 4.6 mm X 100 mm
Catalog No. 502392932 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters BEH-based Glycan chemistry offers highly scalable particle sizes (1.7 μm UPLC, 2.5 μm XP, 3.5 μm HPLC) for diverse application needs. Each batch of BEH Glycan material is QC tested with the 2-AB labeled Waters Glycan Performance standard, ensuring consistent and similar separated glycan profiles. Chromatographers can thus select optimal LC technology for specific released glycan analysis.
  • Available in three highly scalable particle sizes (1.7 μm for UPLC, 2.5 μm XP, and 3.5 μm for HPLC).
  • Each batch of BEH Glycan material is QC tested with 2-AB labeled Glycan Performance standard for consistent, similar separated glycan profiles.
  • Enables chromatographers to choose optimal LC technology for specific released glycan analysis.
Catalog No. 50-239-2932 Supplier Waters Corp Supplier No. 186007269
Waters BEH-based Glycan chemistry offers highly scalable particle sizes (1.7 μm UPLC, 2.5 μm XP, 3.5 μm HPLC) for diverse application needs. Each batch of BEH Glycan material is QC tested with the 2-AB labeled Waters Glycan Performance standard, ensuring consistent and similar separated glycan profiles. Chromatographers can thus select optimal LC technology for specific released glycan analysis.
  • Available in three highly scalable particle sizes (1.7 μm for UPLC, 2.5 μm XP, and 3.5 μm for HPLC).
  • Each batch of BEH Glycan material is QC tested with 2-AB labeled Glycan Performance standard for consistent, similar separated glycan profiles.
  • Enables chromatographers to choose optimal LC technology for specific released glycan analysis.