Learn More
Waters Corp XBridge Glycan BEH Amide Column, 130Å, 2.5μm, 3 mm x 30 mm
Catalog No. 502392742 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
BEH-based Glycan chemistry provides scalable particle sizes for UPLC (1.7 μm) and HPLC (2.5 μm XP, 3.5 μm) applications. Each batch is QC tested with 2-AB labeled Glycan Performance standard, ensuring batch consistency and highly similar separated glycan profiles. Chromatographers can select LC technology to suit their specific released glycan analysis and instrumentation, achieving similar results.
  • Available in three highly scalable particle sizes for UPLC and HPLC applications.
  • Batch quality control tested with 2-AB labeled Glycan Performance standard for consistency.
  • Helps ensure highly similar separated glycan profiles.
  • Allows chromatographers to choose appropriate LC-based technology for specific released glycan analysis.
Catalog No. 50-239-2742 Supplier Waters Corp Supplier No. 186008038
BEH-based Glycan chemistry provides scalable particle sizes for UPLC (1.7 μm) and HPLC (2.5 μm XP, 3.5 μm) applications. Each batch is QC tested with 2-AB labeled Glycan Performance standard, ensuring batch consistency and highly similar separated glycan profiles. Chromatographers can select LC technology to suit their specific released glycan analysis and instrumentation, achieving similar results.
  • Available in three highly scalable particle sizes for UPLC and HPLC applications.
  • Batch quality control tested with 2-AB labeled Glycan Performance standard for consistency.
  • Helps ensure highly similar separated glycan profiles.
  • Allows chromatographers to choose appropriate LC-based technology for specific released glycan analysis.