Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Column, Amide, 2.5 μm, 4.6 mm × 150 mm, 130 Å

Catalog No. 502391219 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This column features BEH-based Glycan chemistry, designed for UPLC and HPLC applications. Each batch undergoes quality control testing with the 2-AB labeled Glycan Performance standard, ensuring consistency and highly similar separated glycan profiles for released glycan analysis.

  • Amide chemistry for hydrophilic interaction (HILIC) separation mode
  • Hybrid particle substrate with spherical shape
  • pH range: 2 to 11
  • Low silanol activity
  • Molecular weight range: 1,000 to 150,000
  • Particle size: 2.5 μm
  • Pore size: 130 Å
  • QC tested for glycan applications
  • Compatible with HPLC systems
  • Inner diameter: 4.6 mm
  • Length: 150 mm
  • Carbon load: 12%

Catalog No. 50-239-1219 Supplier Waters Corp Supplier No. 186007270
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This column features BEH-based Glycan chemistry, designed for UPLC and HPLC applications. Each batch undergoes quality control testing with the 2-AB labeled Glycan Performance standard, ensuring consistency and highly similar separated glycan profiles for released glycan analysis.

  • Amide chemistry for hydrophilic interaction (HILIC) separation mode
  • Hybrid particle substrate with spherical shape
  • pH range: 2 to 11
  • Low silanol activity
  • Molecular weight range: 1,000 to 150,000
  • Particle size: 2.5 μm
  • Pore size: 130 Å
  • QC tested for glycan applications
  • Compatible with HPLC systems
  • Inner diameter: 4.6 mm
  • Length: 150 mm
  • Carbon load: 12%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.