Learn More
Waters Corp XBridge Glycan BEH Amide Column, 2.5 μm, 130 Å, 2.1 x 100 mm
Catalog No. 502391696 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters XBridge Glycan BEH Amide Column is a high-performance HPLC column for robust and reproducible separations of glycans and other highly polar compounds. It uses BEH particle technology and a stable amide phase for HILIC applications, ensuring consistency and transferability between HPLC and UPLC platforms.
  • Ensures consistent glycan profiles and batch uniformity.
  • Stable BEH Amide chemistry for HILIC separations.
  • Effectively separates and retains highly polar compounds.
  • Facilitates separation transfer between HPLC and UPLC.
  • Maintains stability over a broad pH range.
  • Designed for HILIC separations of 2-AB, 2-AA, or RapiFluor-MS™ labeled glycans.
  • Recommended operating pH range: 3 to 8.
  • Recommended operating temperatures: 20°C to 90°C.
Catalog No. 50-239-1696 Supplier Waters Corp Supplier No. 186007264
The Waters XBridge Glycan BEH Amide Column is a high-performance HPLC column for robust and reproducible separations of glycans and other highly polar compounds. It uses BEH particle technology and a stable amide phase for HILIC applications, ensuring consistency and transferability between HPLC and UPLC platforms.
  • Ensures consistent glycan profiles and batch uniformity.
  • Stable BEH Amide chemistry for HILIC separations.
  • Effectively separates and retains highly polar compounds.
  • Facilitates separation transfer between HPLC and UPLC.
  • Maintains stability over a broad pH range.
  • Designed for HILIC separations of 2-AB, 2-AA, or RapiFluor-MS™ labeled glycans.
  • Recommended operating pH range: 3 to 8.
  • Recommended operating temperatures: 20°C to 90°C.