Learn More
Waters Corp Waters XBridge Glycan BEH Amide Column, 130Å, 3.5 μm, 4.6 x 100 mm
Catalog No. 502392272 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These XBridge Glycan BEH Amide Columns are designed for Hydrophilic Interaction Liquid Chromatography (HILIC) of labeled glycans, providing stable and efficient analysis of neutral and charged species.
  • Designed for HILIC separations of 2-aminobenzamide (2-AB), 2-aminobenzoic acid (2-AA), or RapiFluor-MS™ labeled glycans.
  • Separates both neutral and charged labeled glycan species.
  • Features chemical and mechanical stability due to ethylene bridged hybrid particle composition.
  • Scalable particle sizes for UPLC (1.7 μm) and HPLC (2.5 μm XP, 3.5 μm) applications.
  • Recommended operating pH range of 3 to 8.
  • Operates between 20°C to 90°C, enhancing selectivity, lowering solvent viscosity, and increasing mass transfer rates.
Catalog No. 50-239-2272 Supplier Waters Corp Supplier No. 186007274
These XBridge Glycan BEH Amide Columns are designed for Hydrophilic Interaction Liquid Chromatography (HILIC) of labeled glycans, providing stable and efficient analysis of neutral and charged species.
  • Designed for HILIC separations of 2-aminobenzamide (2-AB), 2-aminobenzoic acid (2-AA), or RapiFluor-MS™ labeled glycans.
  • Separates both neutral and charged labeled glycan species.
  • Features chemical and mechanical stability due to ethylene bridged hybrid particle composition.
  • Scalable particle sizes for UPLC (1.7 μm) and HPLC (2.5 μm XP, 3.5 μm) applications.
  • Recommended operating pH range of 3 to 8.
  • Operates between 20°C to 90°C, enhancing selectivity, lowering solvent viscosity, and increasing mass transfer rates.