Learn More
Waters Corp Column, XBridge Glycan BEH Amide, 130 Å, 3.5 μm
Supplier: Waters Corp 186007273
Waters BEH-based Glycan chemistry offers scalable particle sizes for UPLC and HPLC applications. Each batch is QC tested with the 2-AB labeled Glycan Performance standard to ensure consistency and highly similar separated glycan profiles. This enables chromatographers to choose the appropriate LC technology for released glycan analysis.
- Chemistry: Amide
- Separation mode: Hydrophilic interaction (HILIC)
- Particle substrate: Hybrid
- pH range: 2 to 11
- Maximum pressure: 6000 psi (415 Bar)
- Silanol activity: Low
- Molecular weight range: 1000 to 150000
- Particle shape: Spherical
- Particle size: 3.5 μm
- Endfitting type: Standard
- Pore size: 130 Å
- QC tested: Glycan
- System: HPLC
- Particle technology: BEH
- Inner diameter: 4.6 mm
- Length: 50 mm
- Carbon load: 12%
- Application: Glycan
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.