Learn More
Waters Corp Waters Corporation XBridge Glycan BEH Amide, 130Å, 2.5 μm XP, 4.6 x 150 mm
Catalog No. 502392390 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Glycan BEH Amide column is designed for HILIC separations, offering robust analysis of labeled glycan species with excellent resolving power and stability.
  • HILIC separations for 2-AB, 2-AA, or RapiFluor-MS™ labeled glycans.
  • Separates neutral and charged labeled glycan species.
  • Retention based on molecule hydrophobicity.
  • Resolving power from fully porous packing materials.
  • Chemical and mechanical stability from ethylene bridged hybrid particle composition.
  • Scalable UPLC and HPLC BEH Glycan column offerings.
  • Calibrates using Dextran Calibration Ladder Standards (glucose units).
  • Predicts labeled oligosaccharide retention based on hydrophilic contributions.
Catalog No. 50-239-2390 Supplier Waters Corp Supplier No. 186007271
The XBridge Glycan BEH Amide column is designed for HILIC separations, offering robust analysis of labeled glycan species with excellent resolving power and stability.
  • HILIC separations for 2-AB, 2-AA, or RapiFluor-MS™ labeled glycans.
  • Separates neutral and charged labeled glycan species.
  • Retention based on molecule hydrophobicity.
  • Resolving power from fully porous packing materials.
  • Chemical and mechanical stability from ethylene bridged hybrid particle composition.
  • Scalable UPLC and HPLC BEH Glycan column offerings.
  • Calibrates using Dextran Calibration Ladder Standards (glucose units).
  • Predicts labeled oligosaccharide retention based on hydrophilic contributions.