Learn More
Waters Corp XBridge Glycan BEH Amide Method Validation Kit, 130Å, 2.5 μm
Catalog No. 502390861 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • BEH-based Glycan chemistry offerings are available in three highly scalable particle sizes that address UPLC (1.7 μm) and HPLC-based (2.5 μm XP and 3.5 μm) application needs.
  • Each batch of BEH Glycan material is specifically quality control tested with the 2-AB labeled, Glycan Performance standard to help ensure batch to batch consistency as well as highly similar separated glycan profiles.
  • Chromatographers can choose the most appropriate LC-based technology to address their specific released glycan analysis application needs and laboratory instrumentation to ensure highly similar results.
Catalog No. 50-239-0861 Supplier Waters Corp Supplier No. 186007266
  • BEH-based Glycan chemistry offerings are available in three highly scalable particle sizes that address UPLC (1.7 μm) and HPLC-based (2.5 μm XP and 3.5 μm) application needs.
  • Each batch of BEH Glycan material is specifically quality control tested with the 2-AB labeled, Glycan Performance standard to help ensure batch to batch consistency as well as highly similar separated glycan profiles.
  • Chromatographers can choose the most appropriate LC-based technology to address their specific released glycan analysis application needs and laboratory instrumentation to ensure highly similar results.