Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Sentry Guard Cartridge, BEH Amide, 130Å, 3.5 μm, 4.6 mm ID

Catalog No. 502391759 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Waters BEH-based Glycan chemistry offerings are available in three highly scalable particle sizes that address UPLC (1.7 μm) and HPLC-based (2.5 μm XP and 3.5 μm) application needs. Each batch of BEH Glycan material is specifically quality control tested with the 2-AB labeled, Waters Glycan Performance standard to help ensure batch to batch consistency as well as highly similar separated glycan profiles. Chromatographers can choose the most appropriate LC-based technology to address their specific released glycan analysis application needs and laboratory instrumentation.

  • Amide chemistry, hybrid particle substrate, HILIC separation
  • Spherical 3.5 μm particle size, 130 Å pore size
  • pH range: 2 to 11; Max pressure: 6000 psi (415 Bar)
  • Low silanol activity; MW range: 1000 to 150000
  • Glycan QC tested for HPLC systems
  • 4.6 mm ID, 20 mm length; 12% Carbon load

Catalog No. 50-239-1759 Supplier Waters Corp Supplier No. 186007272
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Waters BEH-based Glycan chemistry offerings are available in three highly scalable particle sizes that address UPLC (1.7 μm) and HPLC-based (2.5 μm XP and 3.5 μm) application needs. Each batch of BEH Glycan material is specifically quality control tested with the 2-AB labeled, Waters Glycan Performance standard to help ensure batch to batch consistency as well as highly similar separated glycan profiles. Chromatographers can choose the most appropriate LC-based technology to address their specific released glycan analysis application needs and laboratory instrumentation.

  • Amide chemistry, hybrid particle substrate, HILIC separation
  • Spherical 3.5 μm particle size, 130 Å pore size
  • pH range: 2 to 11; Max pressure: 6000 psi (415 Bar)
  • Low silanol activity; MW range: 1000 to 150000
  • Glycan QC tested for HPLC systems
  • 4.6 mm ID, 20 mm length; 12% Carbon load

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.