Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Sentry Guard Cartridge, XBridge Glycan BEH Amide, 2.5 μm

Catalog No. 502390582 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Sentry Guard Cartridge utilizes BEH-based Glycan chemistry for UPLC and HPLC applications. Each batch undergoes QC testing to ensure batch-to-batch consistency and similar glycan profiles, enabling chromatographers to select suitable LC-based technology for released glycan analysis.

  • Chemistry: Amide
  • Separation mode: HILIC
  • Particle substrate: Hybrid
  • pH range: 2 to 11
  • Not endcapped
  • Low silanol activity
  • MW range: 1,000 to 150,000
  • Spherical particle shape
  • Particle size: 2.5 μm
  • Pore size: 130 Å
  • QC tested for glycan
  • Format: Sentry Guard Cartridge
  • Surface area: 185
  • System: HPLC
  • USP classification: L68
  • Inner diameter: 4.6 mm
  • Length: 20 mm
  • Carbon load: 12%
  • Application: Glycan
  • Units per package: 2

Catalog No. 50-239-0582 Supplier Waters Corp Supplier No. 186007267
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Sentry Guard Cartridge utilizes BEH-based Glycan chemistry for UPLC and HPLC applications. Each batch undergoes QC testing to ensure batch-to-batch consistency and similar glycan profiles, enabling chromatographers to select suitable LC-based technology for released glycan analysis.

  • Chemistry: Amide
  • Separation mode: HILIC
  • Particle substrate: Hybrid
  • pH range: 2 to 11
  • Not endcapped
  • Low silanol activity
  • MW range: 1,000 to 150,000
  • Spherical particle shape
  • Particle size: 2.5 μm
  • Pore size: 130 Å
  • QC tested for glycan
  • Format: Sentry Guard Cartridge
  • Surface area: 185
  • System: HPLC
  • USP classification: L68
  • Inner diameter: 4.6 mm
  • Length: 20 mm
  • Carbon load: 12%
  • Application: Glycan
  • Units per package: 2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.