Learn More
Waters Corp XBridge VanGuard Pre-column, Glycan BEH Amide, 2.5 μm, 130 Å
Catalog No. 502392552 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This VanGuard Pre-column utilizes BEH-based Glycan chemistry, offered in highly scalable particle sizes to suit both UPLC and HPLC applications. Each batch of Glycan material undergoes specific quality control testing with a 2-AB labeled Glycan Performance standard to ensure consistency across batches and highly similar separated glycan profiles. This allows chromatographers to select the most suitable LC-based technology for their released glycan analysis needs.
  • Amide chemistry
  • Hydrophilic Interaction (HILIC) separation mode
  • Hybrid particle substrate
  • pH range from 2 to 11
  • Maximum pressure of 1240 Bar (18000 psi)
  • Spherical particle shape with 2.5 μm particle size
  • Pore size of 130 Å
  • Inner diameter of 2.1 mm and 5 mm length
  • Supplied in a package of 3
Catalog No. 50-239-2552 Supplier Waters Corp Supplier No. 186007262
This VanGuard Pre-column utilizes BEH-based Glycan chemistry, offered in highly scalable particle sizes to suit both UPLC and HPLC applications. Each batch of Glycan material undergoes specific quality control testing with a 2-AB labeled Glycan Performance standard to ensure consistency across batches and highly similar separated glycan profiles. This allows chromatographers to select the most suitable LC-based technology for their released glycan analysis needs.
  • Amide chemistry
  • Hydrophilic Interaction (HILIC) separation mode
  • Hybrid particle substrate
  • pH range from 2 to 11
  • Maximum pressure of 1240 Bar (18000 psi)
  • Spherical particle shape with 2.5 μm particle size
  • Pore size of 130 Å
  • Inner diameter of 2.1 mm and 5 mm length
  • Supplied in a package of 3