Learn More
Waters Corp Waters Corporation Column, 2.5 μm, 130 Å, 2.1 mm × 150 mm
Catalog No. 502037910 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column retains extremely polar compounds like carbohydrates and sugars, separating analytes based on polarity, structure, and pKa. It has a chemically stable, trifunctionally bonded amide phase, stable across a pH range of 2 to 11. Quality control testing with 2-AB labeled Glycan Performance standard ensures batch-to-batch consistency. Based on ethylene bridged hybrid (BEH) particle technology, it offers stability and versatility in HILIC separations, sharing selectivity with similar amide columns for seamless HPLC/UPLC transfer.
  • Chemically stable, trifunctionally bonded amide phase.
  • Stable across a wide pH range of 2 to 11.
  • Batch-to-batch consistency for glycan profiles.
  • Ethylene bridged hybrid (BEH) particle for stability.
  • Versatility in HILIC separations.
  • Seamless transfer between HPLC and UPLC.
  • Withstands high pressures up to 18,000 psi (1240 Bar).
Catalog No. 50-203-7910 Supplier Waters Corp Supplier No. 186007265
This column retains extremely polar compounds like carbohydrates and sugars, separating analytes based on polarity, structure, and pKa. It has a chemically stable, trifunctionally bonded amide phase, stable across a pH range of 2 to 11. Quality control testing with 2-AB labeled Glycan Performance standard ensures batch-to-batch consistency. Based on ethylene bridged hybrid (BEH) particle technology, it offers stability and versatility in HILIC separations, sharing selectivity with similar amide columns for seamless HPLC/UPLC transfer.
  • Chemically stable, trifunctionally bonded amide phase.
  • Stable across a wide pH range of 2 to 11.
  • Batch-to-batch consistency for glycan profiles.
  • Ethylene bridged hybrid (BEH) particle for stability.
  • Versatility in HILIC separations.
  • Seamless transfer between HPLC and UPLC.
  • Withstands high pressures up to 18,000 psi (1240 Bar).