Learn More
Waters Corp Waters Column, XBridge BEH HILIC, 130Å, 3.5 μm, 2.1 mm × 150 mm
Catalog No. 50782413 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH HILIC columns feature unbonded BEH substrate, designed for the retention and separation of highly polar compounds. These columns are optimized and tested for efficient, reproducible separations under HPLC HILIC conditions. They offer greater ruggedness than silica-based HILIC phases, with the BEH particle's chemical stability contributing to longer column lifetimes.
  • Retains and separates highly polar compounds efficiently.
  • Optimized for reproducible HPLC HILIC separations.
  • More rugged than silica-based HILIC phases.
  • BEH particle chemical stability ensures longer column lifetimes.
  • Improved retention for polar analytes via complex mechanisms.
  • Complementary selectivity to reversed-phase columns.
  • Enhanced peak form and longevity.
  • Built on BEH Technology, setting a new standard for HILIC.
  • Maximum pressure: 6000 psi (415 Bar); pH range: 1 to 9.
Catalog No. 50-782-413 Supplier Waters Corp Supplier No. 186004434
XBridge BEH HILIC columns feature unbonded BEH substrate, designed for the retention and separation of highly polar compounds. These columns are optimized and tested for efficient, reproducible separations under HPLC HILIC conditions. They offer greater ruggedness than silica-based HILIC phases, with the BEH particle's chemical stability contributing to longer column lifetimes.
  • Retains and separates highly polar compounds efficiently.
  • Optimized for reproducible HPLC HILIC separations.
  • More rugged than silica-based HILIC phases.
  • BEH particle chemical stability ensures longer column lifetimes.
  • Improved retention for polar analytes via complex mechanisms.
  • Complementary selectivity to reversed-phase columns.
  • Enhanced peak form and longevity.
  • Built on BEH Technology, setting a new standard for HILIC.
  • Maximum pressure: 6000 psi (415 Bar); pH range: 1 to 9.