Learn More
Waters Corp Waters XBridge BEH HILIC Column, 3.5 μm, 2.1 mm ID
Catalog No. 50782411 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These BEH HILIC columns feature unbonded BEH substrate, designed for retention and separation of very polar compounds. Optimized for efficient, reproducible HPLC HILIC separations, they offer extended column lifetimes due to the chemical stability of the BEH particle. This design surpasses silica-based HILIC phases in ruggedness and improves polar analyte retention via HILIC technology, utilizing complex mechanisms and providing superior retention with mobile phases containing over 80 percent acetonitrile.
  • Alternate selectivity and enhanced peak shape compared to reversed-phase columns.
  • Improved retention and column longevity over silica-based HILIC columns.
  • Dependable BEH technology for advanced HILIC separations.
  • Achieves elution reversal and enhanced retentivity for orthogonal polar analyte separations.
Catalog No. 50-782-411 Supplier Waters Corp Supplier No. 186004432
These BEH HILIC columns feature unbonded BEH substrate, designed for retention and separation of very polar compounds. Optimized for efficient, reproducible HPLC HILIC separations, they offer extended column lifetimes due to the chemical stability of the BEH particle. This design surpasses silica-based HILIC phases in ruggedness and improves polar analyte retention via HILIC technology, utilizing complex mechanisms and providing superior retention with mobile phases containing over 80 percent acetonitrile.
  • Alternate selectivity and enhanced peak shape compared to reversed-phase columns.
  • Improved retention and column longevity over silica-based HILIC columns.
  • Dependable BEH technology for advanced HILIC separations.
  • Achieves elution reversal and enhanced retentivity for orthogonal polar analyte separations.