Learn More
Waters Corp Waters XBridge BEH HILIC Column, 130Å, 3.5 μm, 4.6 mm X 50 mm
Catalog No. 50782418 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. These columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions. They are more rugged than silica-based HILIC phases, and the BEH particle's chemical stability results in long column lifetimes.
  • Optimized for polar compounds: designed to retain and separate very polar compounds.
  • Efficient and reproducible separations: optimized and tested for efficient, reproducible separations under HPLC HILIC conditions.
  • Rugged and long-lasting: more rugged than silica-based HILIC phases, leading to long column lifetimes due to chemical stability of the BEH particle.
Catalog No. 50-782-418 Supplier Waters Corp Supplier No. 186004439
XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. These columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions. They are more rugged than silica-based HILIC phases, and the BEH particle's chemical stability results in long column lifetimes.
  • Optimized for polar compounds: designed to retain and separate very polar compounds.
  • Efficient and reproducible separations: optimized and tested for efficient, reproducible separations under HPLC HILIC conditions.
  • Rugged and long-lasting: more rugged than silica-based HILIC phases, leading to long column lifetimes due to chemical stability of the BEH particle.