Learn More
Waters Corp Waters Column, HILIC, 130Å, 3.5 μm, 4.6 mm × 150 mm
Catalog No. 50782420 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions. More rugged than silica based HILIC phases, the BEH particle's chemical stability results in long column lifetimes.
  • Designed to retain and separate very polar compounds.
  • Optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions.
  • More rugged than silica based HILIC phases.
  • BEH particle's chemical stability results in long column lifetimes.
Catalog No. 50-782-420 Supplier Waters Corp Supplier No. 186004441
Add to Cart
Add to Cart
XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions. More rugged than silica based HILIC phases, the BEH particle's chemical stability results in long column lifetimes.
  • Designed to retain and separate very polar compounds.
  • Optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions.
  • More rugged than silica based HILIC phases.
  • BEH particle's chemical stability results in long column lifetimes.