Learn More
Waters Corp XBridge BEH HILIC Column, 130Å, 3.5 µm, 3 mm X 50 mm, 1/pk
Catalog No. 50782414 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions. More rugged than silica based HILIC phases, the BEH particle’s chemical stability results in long column lifetimes.
Catalog No. 50-782-414 Supplier Waters Corp Supplier No. 186004435
XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions. More rugged than silica based HILIC phases, the BEH particle’s chemical stability results in long column lifetimes.