Learn More
Waters Corp XBridge Column, BEH HILIC, 5 μm, 2.1 mm
Catalog No. 50782424 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH HILIC columns contain unbonded BEH substrate, designed for the retention and separation of highly polar compounds. These columns are optimized and tested to deliver efficient, reproducible separations under HPLC HILIC conditions. The BEH particle’s chemical stability contributes to long column lifetimes, making them more rugged than silica-based HILIC phases.
  • Designed to retain and separate very polar compounds.
  • Optimized and tested for efficient, reproducible separations under HPLC HILIC conditions.
  • Chemical stability of BEH particle results in long column lifetimes.
  • More rugged than silica-based HILIC phases.
Catalog No. 50-782-424 Supplier Waters Corp Supplier No. 186004445
XBridge BEH HILIC columns contain unbonded BEH substrate, designed for the retention and separation of highly polar compounds. These columns are optimized and tested to deliver efficient, reproducible separations under HPLC HILIC conditions. The BEH particle’s chemical stability contributes to long column lifetimes, making them more rugged than silica-based HILIC phases.
  • Designed to retain and separate very polar compounds.
  • Optimized and tested for efficient, reproducible separations under HPLC HILIC conditions.
  • Chemical stability of BEH particle results in long column lifetimes.
  • More rugged than silica-based HILIC phases.