Learn More
Waters Corp Waters XBridge BEH HILIC Column, 5 μm, 130 Å, 4.6 mm, 100 mm
Catalog No. 50782431 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These HILIC columns are designed for retaining and separating highly polar compounds, delivering efficient and reproducible separations under HPLC HILIC conditions. Composed of unbonded BEH substrate, they provide greater ruggedness and extended column lifetimes compared to silica-based HILIC phases. They offer improved retention for polar analytes and complementary selectivity to reversed-phase columns.
  • Retains and separates highly polar compounds.
  • Optimized for efficient, reproducible HPLC HILIC separations.
  • Rugged with extended column lifetimes due to BEH particle stability.
  • Offers alternate selectivity and improved retention for polar analytes.
  • Advantages in peak form, retention, and longevity over silica-based HILIC columns.
  • Built on robust and reliable BEH Technology.
  • Operating pH range: 1 to 9.
  • Maximum pressure tolerance: 6,000 psi (415 Bar).
Catalog No. 50-782-431 Supplier Waters Corp Supplier No. 186004452
These HILIC columns are designed for retaining and separating highly polar compounds, delivering efficient and reproducible separations under HPLC HILIC conditions. Composed of unbonded BEH substrate, they provide greater ruggedness and extended column lifetimes compared to silica-based HILIC phases. They offer improved retention for polar analytes and complementary selectivity to reversed-phase columns.
  • Retains and separates highly polar compounds.
  • Optimized for efficient, reproducible HPLC HILIC separations.
  • Rugged with extended column lifetimes due to BEH particle stability.
  • Offers alternate selectivity and improved retention for polar analytes.
  • Advantages in peak form, retention, and longevity over silica-based HILIC columns.
  • Built on robust and reliable BEH Technology.
  • Operating pH range: 1 to 9.
  • Maximum pressure tolerance: 6,000 psi (415 Bar).