Learn More
Waters Corp Waters XBridge Oligonucleotide BEH C18 Column, 2.5 μm, 2.1 x 50 mm
Catalog No. 50782083 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed for fast and efficient lab-scale separations, enhancing throughput for synthetic oligonucleotide samples via ion-pair, reversed-phase chromatography, utilizing hybrid-silica BEH Technology particles functionalized with C18. It offers exceptional resolution and superior column life.
  • Achieves fast, efficient lab-scale separations.
  • Utilizes hybrid-silica BEH Technology particles with C18 chemistry.
  • Offers exceptional sample resolution and extended column life.
  • Enables ion-pair, reversed-phase chromatography for synthetic oligonucleotides.
  • Provides enhanced resolving power for large oligonucleotide sequences.
  • Supports 20°C to 65°C operating temperature for optimized selectivity.
  • Suitable for analyzing modified oligonucleotides.
Catalog No. 50-782-083 Supplier Waters Corp Supplier No. 186003952
This column is designed for fast and efficient lab-scale separations, enhancing throughput for synthetic oligonucleotide samples via ion-pair, reversed-phase chromatography, utilizing hybrid-silica BEH Technology particles functionalized with C18. It offers exceptional resolution and superior column life.
  • Achieves fast, efficient lab-scale separations.
  • Utilizes hybrid-silica BEH Technology particles with C18 chemistry.
  • Offers exceptional sample resolution and extended column life.
  • Enables ion-pair, reversed-phase chromatography for synthetic oligonucleotides.
  • Provides enhanced resolving power for large oligonucleotide sequences.
  • Supports 20°C to 65°C operating temperature for optimized selectivity.
  • Suitable for analyzing modified oligonucleotides.