Learn More
Waters Corp XBridge Peptide BEH C18 OBD Prep Column, 130Å, 5 μm, 10 mm X 150 mm
Catalog No. 502391692 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed for peptide separations, featuring BEH C18 chemistry with 130Å pore and 5 μm particle size. It offers a wide pH range (1-12), excellent low pH stability, and minimal column bleed, ideal for proteomics, protein characterization, and peptide synthesis. Its Optimum Bed Density (OBD) design simplifies lab-scale separations, enhancing throughput and enabling scalability from UPLC/HPLC screening to preparative purification.
  • Optimized and QC tested for peptide separations.
  • BEH particle provides wide pH range (1-12) and superior low pH stability with ultra-low column bleed.
  • OBD preparative column simplifies lab-scale separations, enhancing throughput and scalability.
  • Manufactured in a cGMP, ISO 9002 accredited plant for consistent quality and stability.
  • Ensures good peak shape and high efficiency; each batch undergoes peptide separation testing.
Catalog No. 50-239-1692 Supplier Waters Corp Supplier No. 186008188
This column is designed for peptide separations, featuring BEH C18 chemistry with 130Å pore and 5 μm particle size. It offers a wide pH range (1-12), excellent low pH stability, and minimal column bleed, ideal for proteomics, protein characterization, and peptide synthesis. Its Optimum Bed Density (OBD) design simplifies lab-scale separations, enhancing throughput and enabling scalability from UPLC/HPLC screening to preparative purification.
  • Optimized and QC tested for peptide separations.
  • BEH particle provides wide pH range (1-12) and superior low pH stability with ultra-low column bleed.
  • OBD preparative column simplifies lab-scale separations, enhancing throughput and scalability.
  • Manufactured in a cGMP, ISO 9002 accredited plant for consistent quality and stability.
  • Ensures good peak shape and high efficiency; each batch undergoes peptide separation testing.