Learn More
Waters Corp XBridge Peptide BEH C18 OBD Column, 300 Å, 10 μm, 10 x 50 mm
Catalog No. 502391437 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Peptide BEH C18, 300 Å is an OBD preparative column designed for peptide analysis. It utilizes BEH (Ethylene-Bridged Hybrid) particles and is QC tested with a complex peptide mixture to ensure consistent column-to-column performance.
  • OBD Column type
  • XBridge Peptide BEH C18 chemistry
  • Pore size: 300 Å
  • Particle size: 10 μm
  • Dimensions: 10 x 50 mm
  • QC tested with a complex peptide mixture
Catalog No. 50-239-1437 Supplier Waters Corp Supplier No. 186008198
The XBridge Peptide BEH C18, 300 Å is an OBD preparative column designed for peptide analysis. It utilizes BEH (Ethylene-Bridged Hybrid) particles and is QC tested with a complex peptide mixture to ensure consistent column-to-column performance.
  • OBD Column type
  • XBridge Peptide BEH C18 chemistry
  • Pore size: 300 Å
  • Particle size: 10 μm
  • Dimensions: 10 x 50 mm
  • QC tested with a complex peptide mixture