Learn More
Waters Corp XBridge Peptide BEH C18 Preparative Column, 300 Å, 5 μm
Catalog No. 502391438 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These preparative columns are engineered for peptide isolations, featuring a trifunctional C18 ligand bonded to Ethylene-Bridged Hybrid (BEH) particles. They are designed to offer different separation selectivities for a broad spectrum of peptides and small proteins.
  • Outstanding peak capacity and superior peak shape with trifluoroacetic acid (TFA) or formic acid (FA) ion pair eluents.
  • Reproducible preparative column performance, ensured by rigorous quality control testing with a complex protein digest.
  • Suitable for peptide isolations, providing selectivity for a wide range of peptides and small proteins.
  • Robust stability across a wide pH range and at high temperatures, due to Ethylene-Bridged Hybrid Technology.
Catalog No. 50-239-1438 Supplier Waters Corp Supplier No. 186008189
Add to Cart
Add to Cart
These preparative columns are engineered for peptide isolations, featuring a trifunctional C18 ligand bonded to Ethylene-Bridged Hybrid (BEH) particles. They are designed to offer different separation selectivities for a broad spectrum of peptides and small proteins.
  • Outstanding peak capacity and superior peak shape with trifluoroacetic acid (TFA) or formic acid (FA) ion pair eluents.
  • Reproducible preparative column performance, ensured by rigorous quality control testing with a complex protein digest.
  • Suitable for peptide isolations, providing selectivity for a wide range of peptides and small proteins.
  • Robust stability across a wide pH range and at high temperatures, due to Ethylene-Bridged Hybrid Technology.