Learn More
Waters Corp Waters OBD Column, XBridge Peptide BEH C18, 300 Å, 10 μm
Catalog No. 502392554 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This OBD Column features an XBridge Peptide BEH C18, 300 Å chemistry with a particle size of 10 μm and dimensions of 10 x 150 mm. It is designed for high-performance separations of peptides and small proteins.
  • Trifunctional C18 ligand, fully end-capped, and bonded to ethylene-bridged hybrid (BEH) particles.
  • Offers outstanding peak capacity and superior peak shape in trifluoroacetic acid (TFA) or formic acid (FA) ion pair eluents compared to 100% silica based C18 columns.
  • The 300 Å pore size provides separation selectivity for a wide range of peptides and small proteins.
  • Each synthesized batch is specifically QC tested with a complex protein digest to ensure reproducible prep column performance.
  • Specifically QC tested with a complex peptide mixture to ensure column-to-column performance consistency.
Catalog No. 50-239-2554 Supplier Waters Corp Supplier No. 186008196
Add to Cart
Add to Cart
This OBD Column features an XBridge Peptide BEH C18, 300 Å chemistry with a particle size of 10 μm and dimensions of 10 x 150 mm. It is designed for high-performance separations of peptides and small proteins.
  • Trifunctional C18 ligand, fully end-capped, and bonded to ethylene-bridged hybrid (BEH) particles.
  • Offers outstanding peak capacity and superior peak shape in trifluoroacetic acid (TFA) or formic acid (FA) ion pair eluents compared to 100% silica based C18 columns.
  • The 300 Å pore size provides separation selectivity for a wide range of peptides and small proteins.
  • Each synthesized batch is specifically QC tested with a complex protein digest to ensure reproducible prep column performance.
  • Specifically QC tested with a complex peptide mixture to ensure column-to-column performance consistency.