Learn More
Waters Corp XBridge Peptide BEH C18 Column, 300Å, 10 μm, 50 mm x 250 mm
Catalog No. 502392720 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Peptide BEH C18 OBD Prep Column is designed for peptide separations, optimized and QC tested with cytochrome c digest standards. It utilizes a wide-pore (300Å) trifunctionally bonded BEH particle, providing a wide usable pH range, superior low pH stability, and ultra-low column bleed. Ideal for assaying samples with large peptides in proteomics, protein characterization, and peptide synthesis.
  • Optimized for peptide separations and QC tested with cytochrome c digest standards.
  • Features a wide-pore (300Å) trifunctionally bonded BEH particle.
  • Offers a wide usable pH range (1 to 12) with superior low pH stability and ultra-low column bleed.
  • Ideal for assaying large peptides in proteomics, protein characterization, and peptide synthesis.
  • Enables direct scaling from UPLC, UHPLC, or HPLC screening to lab-scale purification.
Catalog No. 50-239-2720 Supplier Waters Corp Supplier No. 186009428
Add to Cart
Add to Cart
Request a Quote
The XBridge Peptide BEH C18 OBD Prep Column is designed for peptide separations, optimized and QC tested with cytochrome c digest standards. It utilizes a wide-pore (300Å) trifunctionally bonded BEH particle, providing a wide usable pH range, superior low pH stability, and ultra-low column bleed. Ideal for assaying samples with large peptides in proteomics, protein characterization, and peptide synthesis.
  • Optimized for peptide separations and QC tested with cytochrome c digest standards.
  • Features a wide-pore (300Å) trifunctionally bonded BEH particle.
  • Offers a wide usable pH range (1 to 12) with superior low pH stability and ultra-low column bleed.
  • Ideal for assaying large peptides in proteomics, protein characterization, and peptide synthesis.
  • Enables direct scaling from UPLC, UHPLC, or HPLC screening to lab-scale purification.