Learn More
Waters Corp Peptide Column, BEH C18, 130Å, 10 μm, 10 x 100 mm
Supplier:  Waters Corp 186008195
Encompass_Preferred
This column is designed for peptide separations, utilizing Ethylene-Bridged Hybrid (BEH) particles with a C18 ligand.
  • Outstanding peak shape and capacity in TFA or FA containing eluents compared to 100% silica-based C18 columns.
  • BEH particles are stable from pH 1 to 11 and up to 80°C, expanding method development capabilities for peptide separation selectivities.
  • The 130Å pore size, along with a 300Å pore size, provides different separation selectivities for peptides and small proteins.
  • Each batch of synthesized Peptide BEH C18 130Å particles is rigorously tested to separate a complex protein digest using a gradient separation with well-defined criteria.
  • Each manufactured column is tested and must exceed established packed column efficiency values for consistent batch-to-batch and column-to-column performance.
Catalog No. 50-239-1426