Learn More
Waters Corp Waters Prep Column, C18, BEH, 300Å, 10 μm
Supplier: Waters Corp 186008201
The XBridge BEH300 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. Featuring a wide-pore (300Å) trifunctionally bonded BEH particle, they offer a wide usable pH range, superior low pH stability, and ultra-low column bleed for proteomics, protein characterization, and peptide synthesis applications.
- C18 chemistry.
- Hybrid particle substrate.
- 300Å pore size.
- 10 μm particle size.
- 10 mm × 250 mm dimensions.
- pH range 1 to 12.
- Maximum pressure 6000 psi (415 bar).
- Molecular weight range 1,000 to 15,000.
- Spherical particle shape.
- OBD prep column format.
- For HPLC systems.
- BEH particle technology ensures stable bonded phases.
- Negligible retention loss in low-pH stability tests.
- Physical stability via Optimum Bed Density (OBD) Technology.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.