Learn More
Waters Corp Prep Guard Cartridge, C18, 300Å, 5 μm, 30 mm x 10 mm
Supplier:  Waters Corp 186006883
Encompass_Preferred
This Prep Guard Cartridge is designed to be used with a 30 mm x 10 mm Prep Guard Holder to protect both HPLC and SFC 30 mm ID Prep OBD™ columns. Placing this guard assembly before the prep column helps extend column lifetimes and reduce purification costs.
  • Protects both HPLC and SFC 30 mm ID Prep OBD™ columns
  • Provides longer column lifetimes
  • Reduces purification costs
Catalog No. 50-239-1765