Learn More
Waters Corp Column, XBridge Peptide BEH C18 XP, 300Å, 2.5 μm, 3 mm × 100 mm
Catalog No. 502391241 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Peptide BEH C18 XP Column, 300Å, 2.5 μm, 3mm × 100 mm Columns are optimized and QC tested for peptide separations. The 300Å pore size trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed for especially large peptides in proteomic, peptide mapping, and synthetic peptide applications. Each batch is specifically QC tested using a gradient separation of a tryptic digest of cytochrome c before being QC accepted as a Peptide separation column.
  • Optimized and QC tested for peptide separations.
  • 300Å pore size trifunctionally bonded BEH particle.
  • Widest usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed for especially large peptides.
  • Each batch QC tested using a gradient separation of a tryptic digest of cytochrome c.
Catalog No. 50-239-1241 Supplier Waters Corp Supplier No. 186009072
The XBridge Peptide BEH C18 XP Column, 300Å, 2.5 μm, 3mm × 100 mm Columns are optimized and QC tested for peptide separations. The 300Å pore size trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed for especially large peptides in proteomic, peptide mapping, and synthetic peptide applications. Each batch is specifically QC tested using a gradient separation of a tryptic digest of cytochrome c before being QC accepted as a Peptide separation column.
  • Optimized and QC tested for peptide separations.
  • 300Å pore size trifunctionally bonded BEH particle.
  • Widest usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed for especially large peptides.
  • Each batch QC tested using a gradient separation of a tryptic digest of cytochrome c.