Learn More
Waters Corp XBridge Peptide BEH C18 XP Column, 300Å, 2.5 μm, 4.6 mm × 50 mm
Catalog No. 502392531 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Peptide BEH C18 XP Column is optimized and QC tested for peptide separations. Its 300Å pore size trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed, making it suitable for large peptides in proteomic, peptide mapping, and synthetic peptide applications. Each batch undergoes specific QC testing for peptide separation acceptance.
  • Effective for peptide separations.
  • Wide usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed.
  • Each batch QC tested.
  • Suitable for proteomic, peptide mapping, and synthetic peptide applications.
  • Features BEH (Bridged Ethyl Hybrid) particle technology.
  • USP classification L1.
  • Compatible with HPLC and UHPLC systems.
Catalog No. 50-239-2531 Supplier Waters Corp Supplier No. 186009074
Add to Cart
Add to Cart
The XBridge Peptide BEH C18 XP Column is optimized and QC tested for peptide separations. Its 300Å pore size trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed, making it suitable for large peptides in proteomic, peptide mapping, and synthetic peptide applications. Each batch undergoes specific QC testing for peptide separation acceptance.
  • Effective for peptide separations.
  • Wide usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed.
  • Each batch QC tested.
  • Suitable for proteomic, peptide mapping, and synthetic peptide applications.
  • Features BEH (Bridged Ethyl Hybrid) particle technology.
  • USP classification L1.
  • Compatible with HPLC and UHPLC systems.