Learn More
Waters Corp XBridge Peptide BEH C18 XP Column, 300Å, 2.5 μm, 2.1 mm
Catalog No. 502392539 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Peptide BEH C18 XP Column is specifically optimized and QC tested for peptide separations, making it ideal for proteomic, peptide mapping, and synthetic peptide applications. Its 300Å pore size BEH particle design provides a wide usable pH range, superior low pH stability, and ultra-low column bleed.
  • Optimized and QC tested for peptide separations.
  • Features 300Å pore size BEH particle for wide pH range and low column bleed.
  • Suitable for large peptides in proteomic, peptide mapping, and synthetic peptide applications.
  • 2.1 mm ID columns recommended for UPLC or UHPLC systems, especially LC-MS methods.
  • Each batch QC tested using a gradient separation of tryptic digest of cytochrome c.
  • Suggested for peptide separations and analysis.
Catalog No. 50-239-2539 Supplier Waters Corp Supplier No. 186009070
The XBridge Peptide BEH C18 XP Column is specifically optimized and QC tested for peptide separations, making it ideal for proteomic, peptide mapping, and synthetic peptide applications. Its 300Å pore size BEH particle design provides a wide usable pH range, superior low pH stability, and ultra-low column bleed.
  • Optimized and QC tested for peptide separations.
  • Features 300Å pore size BEH particle for wide pH range and low column bleed.
  • Suitable for large peptides in proteomic, peptide mapping, and synthetic peptide applications.
  • 2.1 mm ID columns recommended for UPLC or UHPLC systems, especially LC-MS methods.
  • Each batch QC tested using a gradient separation of tryptic digest of cytochrome c.
  • Suggested for peptide separations and analysis.